missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Indoleamine 2,3-dioxygenase/IDO Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC6030R-100UG
This item is not returnable.
View return policy
Description
Indoleamine 2,3-dioxygenase/IDO Monoclonal specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Indoleamine 2,3-dioxygenase/IDO | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P14902 | |
| IDO1 | |
| E. coli-derived recombinant human Indoleamine 2,3-dioxygenase/IDO Ala2-Gly403 Accession # P14902 | |
| RUO | |
| 3620 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 700838 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| 3dioxygenase, EC 1.13.11.52, IDO, IDO1, IDOIDO-1, INDO, INDOindole 2,3-dioxygenase, Indoleamine 2, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Indoleamine 2,3-dioxygenase/IDO in direct ELISA. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction