missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Inhibin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB338-SP
This item is not returnable.
View return policy
Description
Inhibin Monoclonal specifically detects Inhibin in Human samples. It is validated for ELISA.
Specifications
| Inhibin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| INHA, INHB, Inhibin A | |
| Chinese hamster ovary cell line CHO-derived recombinant human Inhibin alpha chain, Accession No. P08476 | |
| 25 μg | |
| Detects human Inhibin alpha chain in direct ELISAs. In sandwich immunoassays, this antibody captures the human Inhibin alpha chain and detects either human Inhibin A or human Inhibin B based on the detection antibody. | |
| Human | |
| Supernatant |
| ELISA | |
| 130408R | |
| ELISA | |
| P05111 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction