missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 8 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6194R-100UG
This item is not returnable.
View return policy
Description
Integrin alpha 8 Monoclonal specifically detects Integrin alpha 8 in Human samples. It is validated for Flow Cytometry.
Specifications
| Integrin alpha 8 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P53708 | |
| ITGA8 | |
| E. coli-derived recombinant human Integrin alpha 8 Glu610-Leu800 Accession # P53708 | |
| RUO | |
| 8516 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 481709 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| integrin alpha-8, integrin, alpha 8, ITGA8 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Integrin alpha 8 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction