missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin beta 3/CD61 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2266T-100UG
This item is not returnable.
View return policy
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Flow Cytometry.
Specifications
| Integrin beta 3/CD61 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P05106 | |
| ITGB3 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 3/CD61 Gly27-Asp718 Accession # P05106 | |
| RUO | |
| 3690 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 256809 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD61, CD61 antigen, GP3A, GP3Aplatelet glycoprotein IIIa, GPIIIA, GPIIIaintegrin beta-3, INGRB3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, ITGB3, Platelet membrane glycoprotein IIIa | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Integrin beta 3/CD61 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Integrin beta 1, beta 2, recombinant mouse Integrin beta 4, or beta 6 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction