missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin beta 7 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4669N-100UG
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Human samples. It is validated for Flow Cytometry.
Specifications
| Integrin beta 7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P26010 | |
| ITGB7 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 7 long isoform Glu20-His723 Accession # P26010 | |
| RUO | |
| 3695 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 473207 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7, ITGB7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Integrin beta 7 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Integrin beta 1, beta 2, beta 3, beta 4, beta 5, beta 6, beta 8, recombinant mouse Integrin beta 4, or beta 7 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction