Learn More
Invitrogen™ Human IL-2 Recombinant Protein
Description
Mature human IL-2 was expressed in Pichia pastoris, X-33. GenBank Accession # NP_000577, amino acids 21 - 153. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: APTSSSTKKT. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity was determined by measuring the proliferation of mouse HEK-Blue cells. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 1 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.
Specifications
Specifications
| Accession Number | P60568 |
| For Use With (Application) | Control, ELISA, Immunoprecipitation, Western Blot |
| Formulation | PBS with no preservative |
| Gene ID (Entrez) | 3558 |
| Molecular Weight (g/mol) | 15 kDa |
| Name | Human IL-2 |
| Quantity | 20 μg |
| Source | Yeast |
| Regulatory Status | RUO |
| Gene Alias | aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.