Learn More
Human Irisin/FNDC5 Antibody, Novus Biologicals™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB94201-SP
 View more versions of this product
Product Code. 18745885
  • € 176.00 / 25µg
Description
Irisin/FNDC5 Monoclonal antibody specifically detects Irisin/FNDC5 in nan samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Irisin/FNDC5
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2
Synthetic peptide covering the C-term of the Irisin peptide sequences of the FNDC5 gene.,, Glu97-Glu143, Accession # Q8NAU1
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Purified
Immunocytochemistry
1056111
Immunocytochemistry 8-25 ug/mL
Q8NAU1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
252995
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a