Learn More
R&D Systems™ Human ITIH5 (NP_085046.5) VersaClone cDNA
Specifications
Specifications
| Gene | ITIH5 |
| Accession Number | NP_085046.5 |
| Gene ID (Entrez) | 80760 |
| Gene Alias | inter-alpha (globulin) inhibitor H5, inter-alpha-inhibitor heavy chain 5, inter-alpha-trypsin inhibitor heavy chain H5, ITI heavy chain H5, ITIH5 inter-alpha-trypsin inhibitor heavy chain family, member 5, ITI-HC5, KIAA1953, MGC10848, PP14776 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |