missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ki67/MKI67 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC7617R-100UG
This item is not returnable.
View return policy
Description
Ki67/MKI67 Monoclonal specifically detects Ki67/MKI67 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Ki67/MKI67 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, MKI67, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105, TSG126 | |
| Human Ki-67/MKI67 synthetic peptide Accession # P46013 | |
| 100 μg | |
| 4288 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 1297A | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P46013 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Ki-67/MKI67 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction