missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DS1/CD158h PE-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems FAB8887P-100
This item is not returnable.
View return policy
Description
KIR2DS1/CD158h Monoclonal specifically detects KIR2DS1/CD158h in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR2DS1/CD158h | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| killer cell immunoglobulin-like receptor 2DS1, CD158a, CD158H, EB6ActI, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1, p50.1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3806 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1127B | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q14954 | |
| KIR2DS1 | |
| HEK293 human embryonic kidney cell line transfected with human KIR2DS1/CD158h Met1-His245 Accession No. Q14954 | |
| 100 Tests | |
| Primary | |
| Detects human KIR2DS1/CD158h in flow cytometry. Clone 1127B recognizes KIR2DS1 and some alleles of KIR2DL1, both members of the killer cell immunoglobulin-like receptor (KIR) family. Because Clone 1127B displays partial cross-reactivity with KIR2DL1, co-staining with Mouse Anti-Human KIR2DL1 Clone 143211 (Catalog # FAB1844A) is recommended. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction