Learn More
Human KIR2DS1/CD158h PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody has been used in 1 publication
Brand:  R&D Systems FAB8887P-025
 View more versions of this product
Product Code. 18732018
  • ONLINE EXCLUSIVE PRICE
    € 262.00
    € 247.80 / 25 tests
    Save € 14.20
    5% Off
Description
KIR2DS1/CD158h Monoclonal specifically detects KIR2DS1/CD158h in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KIR2DS1/CD158h
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
killer cell immunoglobulin-like receptor 2DS1, CD158a, CD158H, EB6ActI, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1, p50.1
Rabbit
Protein A or G purified from cell culture supernatant
RUO
3806
Human
Purified
Flow Cytometry
1127B
Flow Cytometry 10 uL/10^6 cells
Q14954
KIR2DS1
HEK293 human embryonic kidney cell line transfected with human KIR2DS1/CD158h Met1-His245 Accession # Q14954
25 Tests
Primary
Detects human KIR2DS1/CD158h in flow cytometry. Clone 1127B recognizes KIR2DS1 and some alleles of KIR2DL1, both members of the killer cell immunoglobulin-like receptor (KIR) family. Because Clone 1127B displays partial cross-reactivity with KIR2DL1, co-staining with Mouse Anti-Human KIR2DL1 Clone 143211 (Catalog # FAB1844A) is recommended.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.