missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DS4/CD158i Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1847T-100UG
This item is not returnable.
View return policy
Description
KIR2DS4/CD158i Monoclonal specifically detects KIR2DS4/CD158i in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR2DS4/CD158i | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P43632 | |
| KIR2DS4 | |
| BaF3 mouse pro-B cell line transfected with human KIR2DS4 Accession # P43632 | |
| RUO | |
| 3809 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 179315 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD158 antigen-like family member I, CD158I, CD158i antigen, cl-39, killer cell immunoglobulin-like receptor 2DS4, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail4,nkat8, killer inhibitory receptor 4-1-2, KIR antigen 2DS4, KKA3KIR1D, MGC120019, MGC125315, MGC125317, MHC class I NK cell receptor, natural killer cell inhibitory receptor, Natural killer-associated transcript 8, NKAT-8, NKAT8KIR412, P58 natural killer cell receptor clones CL-39/CL-17, p58 NK receptor CL-39/CL-17 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects KIR2DS4/CD158i in Western blots. In flow cytometry, this antibody stains cells transfected with KIR2DS4/CD158i. Does not stain cells transfected with KIR2DL1, 2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 3DL1, 3DL2, or 3DS1. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction