missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR3DL3/CD158z Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB8919R-025
This item is not returnable.
View return policy
Description
KIR3DL3/CD158z Monoclonal specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR3DL3/CD158z | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| KIR3DL3 | |
| Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z Met1-Leu322 Accession # Q8N743 | |
| 25 Tests | |
| Primary | |
| Detects human KIR3DL3/CD158z in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
| Flow Cytometry | |
| 1136B | |
| Flow Cytometry 5 uL/10^6 cells | |
| Q8N743 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 115653 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction