Learn More
R&D Systems™ Human KLHL13 (NP_277030.2) VersaClone cDNA
Specifications
Specifications
| Gene | KLHL13 |
| Gene ID (Entrez) | 90293 |
| Gene Alias | BTB and kelch domain containing 2, BTB and kelch domain-containing protein 2, kelch-like 13 (Drosophila), kelch-like protein 13, KIAA1309, MGC74791 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |