missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KLRA1 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
KLRA1 Synthetic Peptide, PEP-0792, from Invitrogen.
Ly49A is a gene that encodes a protein expressed on the surface of natural killer cells, which is involved in recognizing and eliminating virally infected or cancerous cells. It is located on chromosome 6 and is part of a larger family of genes that encode for similar receptors. The Ly49A protein interacts with a variety of ligands, including certain viruses and tumor cells, and is involved in regulating the activation and function of natural killer cells. Mutations or variations in the Ly49A gene have been associated with various diseases and conditions, including autoimmune disorders and cancer.
Specifications
Specifications
| Gene ID (Entrez) | 10748 |
| Species | Human |
| Purification Method | purified |
| Common Name | Human KLRA1 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction