missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kynureninase Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4887-SP
This item is not returnable.
View return policy
Description
Kynureninase Polyclonal specifically detects Kynureninase in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Kynureninase | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.7.1.3, KYNU, kynureninase, kynureninase (L-kynurenine hydrolase), L-kynurenine hydrolase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8942 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q16719 | |
| KYNU | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Kynureninase Met1-Asn465 Accession # Q16719 | |
| 25 μg | |
| Primary | |
| Detects human Kynureninase in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction