Learn More
Human L-Selectin/CD62L Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB9787R-100
Product Code. 18771419
  • € 369.00 / 100 tests
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
L-Selectin/CD62L
Monoclonal
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide.
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Binds to CHO cells expressing the human L-Selectin/CD62L protein. It does not recognize CHO cells expressing either the human E-Selectin or human P-Selectin.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
4G8
Flow Cytometry 5 uL/10^6 cells
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1
Recombinant human L-Selectin/CD62L Extracellular domain
100 Tests
6402
Human
Supernatant