missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human L1CAM Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF277-SP
This item is not returnable.
View return policy
Description
L1CAM Polyclonal specifically detects L1CAM in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| L1CAM | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antigen identified by monoclonal R1, CAML1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3897 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.25 - 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| CAA42508 | |
| L1CAM | |
| Mouse myeloma cell line NS0-derived recombinant human L1CAM Ile20-Glu1120 Accession # CAA42508 | |
| 25 μg | |
| Primary | |
| Detects human L1CAM in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) ICAM-2 and rhICAM-3 is observed and less than 1% cross-reactvity with rhCD31 and recombinant mouse VCAM-1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction