missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lactate Dehydrogenase B/LDHB Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC9205R-100UG
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase B Monoclonal specifically detects Lactate Dehydrogenase B in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Lactate Dehydrogenase B | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| EC 1.1.1, EC 1.1.1.27, Epididymis Secretory Protein Li 281, HEL-S-281, lactate dehydrogenase B, LDH heart subunit, LDHB, LDH-B, LDHBD, LDH-H, L-lactate dehydrogenase B chain, Renal Carcinoma Antigen NY-REN-46, TRG-5 | |
| E. coli-derived recombinant human LDHB Met1-Val93 Accession # P07195 | |
| RUO | |
| 3945 | |
| Human | |
| IgG |
| Flow Cytometry | |
| 2057D | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P07195 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Detects human LDHB in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction