missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAIR1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2664U-100UG
This item is not returnable.
View return policy
Description
LAIR1 Monoclonal specifically detects LAIR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| LAIR1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6GTX8 | |
| LAIR1 | |
| Mouse myeloma cell line NS0-derived recombinant human LAIR1 isoform 1 Gln22-His163 Accession # Q6GTX8 | |
| 100 μg | |
| Primary | |
| Detects human LAIR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LAIR2 or recombinant mouse LAIR1 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
| Flow Cytometry | |
| 342219 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD305, CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3903 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction