missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Laminin alpha 1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4187U-100UG
This item is not returnable.
View return policy
Description
Laminin alpha 1 Monoclonal specifically detects Laminin alpha 1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Laminin alpha 1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| LAMA, LAMA1, LAMA1 laminin, alpha 1, S-LAM-alpha | |
| E. coli-derived recombinant human Laminin alpha 1, Leu22-Met269, Accession # P25391 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 775016 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P25391 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284217 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction