Learn More
Human Laminin alpha 4 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB7340-SP
Product Code. 18787769
  • € 208.00 / 25µg
Description
Laminin alpha 4 Monoclonal specifically detects Laminin alpha 4 in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laminin alpha 4
Monoclonal
LYOPH
Immunocytochemistry 8-25 ug/mL
Q16363
LAMA4
Chinese hamster ovary cell line CHO-derived recombinant human Laminin alpha 4 Arg826-Ser1816 Accession # Q16363
25 μg
Primary
Detects human Laminin alpha 4 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Laminin alpha 4 or recombinant human Laminin alpha 3 is observed.
Human
Purified
Immunocytochemistry
839084
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
DKFZp686D23145, LAMA3LAMA4*-1, LAMA4, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3910
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.