missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAMP-1/CD107a APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC4800A
This item is not returnable.
View return policy
Description
LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Human samples. It is validated for Flow Cytometry, Intracellular Staining by Flow Cytometry.
Specifications
| LAMP-1/CD107a | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP1, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3916 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 508921 | |
| Flow Cytometry 10 uL/10^6 cells, Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P11279 | |
| LAMP1 | |
| Mouse myeloma cell line NS0-derived recombinant human LAMP-1/CD107a Ala28-Asn380 Accession # P11279 | |
| 100 Tests | |
| Primary | |
| Detects human LAMP-1/CD107a in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction