missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAP (TGF-beta 1) Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2463U-100UG
This item is not returnable.
View return policy
Description
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Flow Cytometry.
Specifications
| LAP (TGF-beta 1) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P01137 | |
| TGFB1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-beta 1) Leu30-Arg278 (Cys33Ser) Accession # P01137 | |
| RUO | |
| 7040 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 27232 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CED, DPD1, LAP, LAP (TGFbeta 1), TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LAP (TGF-beta 1) in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) TGF-beta 1, rhTGF-beta 2, rhTGF-beta 1.2, rhTGF-beta 3, or rhTGF-alpha . | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction