missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAT Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB63341
This item is not returnable.
View return policy
Description
LAT Monoclonal specifically detects LAT in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| LAT | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| O43561 | |
| LAT | |
| E. coli-derived recombinant human LAT His30-Gln113 Accession # O43561 | |
| 100 μg | |
| Primary | |
| Detects human LAT in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 661002 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27040 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction