missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LBP Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF870-SP
This item is not returnable.
View return policy
Description
LBP Polyclonal specifically detects LBP in Human samples. It is validated for Western Blot.
Specifications
| LBP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3929 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AAD21962 | |
| LBP | |
| Mouse myeloma cell line NS0-derived recombinant human LBP Ala26-Val481 Accession # AAD21962 | |
| 25 μg | |
| Primary | |
| Detects human LBP in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse LBP is observed and less than 1% cross-reactivity with recombinant human BPI is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction