missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Legumain/Asparaginyl Endopeptidase Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2199-SP
This item is not returnable.
View return policy
Description
Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Legumain/Asparaginyl Endopeptidase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q99538 | |
| LGMN | |
| NS0-derived recombinant human Legumain/Asparaginyl Endopeptidase Ile18-Tyr433 Accession # Q99538 | |
| 25 μg | |
| Primary | |
| Detects human Legumain/Asparaginyl Endopeptidase (aa18-433) in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Legumain is observed. Recognizes the pro form of human Legumain and does not recognize the mature form (aa 18-323). | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 312114 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5641 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction