missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Leptin R Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF389
This item is not returnable.
View return policy
Description
Leptin R Polyclonal specifically detects Leptin R in Human samples. It is validated for Western Blot.
Specifications
| Leptin R | |
| Polyclonal | |
| Biotin | |
| P48357 | |
| LEPR | |
| Mouse myeloma cell line NS0-derived recombinant human Leptin R Thr20-Asp839 Accession # P48357 | |
| 50 μg | |
| Primary | |
| Detects human Leptin R in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Leptin R is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3953 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction