missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lgr4/GPR48 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7750S-100UG
This item is not returnable.
View return policy
Description
Lgr4/GPR48 Monoclonal specifically detects Lgr4/GPR48 in Human samples. It is validated for Flow Cytometry.
Specifications
| Lgr4/GPR48 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BXB1 | |
| LGR4 | |
| NS0 mouse myeloma cell line transfected with human Lgr4/GPR48 Met1-Asp951 Accession # Q9BXB1 | |
| RUO | |
| 55366 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 852229 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| GPR48, GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Lgr4/GPR48 in Lgr4-transfected cell-based ELISA. Stains human Lgr4/GPR48 transfected but not irrelevant transfected cells in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction