missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lgr5/GPR49 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB80781-100
This item is not returnable.
View return policy
Description
Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Lgr5/GPR49 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
| Chinese hamster ovary cell line, CHO derived recombinant human Lgr5/GPR-49, Met1-Ile560, Accession # O75473 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 750835 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| O75473 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8549 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction