missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LIF Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2501V-100UG
This item is not returnable.
View return policy
Description
LIF Monoclonal specifically detects LIF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| LIF | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LIF in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse LIF, recombinant human (rh) CLC, rhCNTF, rhCardiotrophin-1, rhIL-6, rhIL-11, or rhOSM is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 9808 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| LIF | |
| E. coli-derived recombinant human LIF | |
| RUO | |
| 3976 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction