Learn More
Human LIF Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB2502-100
 View more versions of this product
Product Code. 18796025
  • ONLINE EXCLUSIVE PRICE
    € 560.00
    € 529.20 / 100µg
    Save € 30.80
    5% Off
Description
LIF Monoclonal antibody specifically detects LIF in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LIF
Monoclonal
Unconjugated
P15018
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3976
Human
Purified
ELISA
945842
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Human embryonic kidney cell, HEK293-derived human LIF, Met1-Phe202, Accession # P15018
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG2b