missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LIFR alpha Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB249V-100UG
This item is not returnable.
View return policy
Description
LIFR alpha Monoclonal specifically detects LIFR alpha in Human samples. It is validated for Flow Cytometry.
Specifications
| LIFR alpha | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P42702 | |
| LIFR | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LIF R alpha Gln45-Ser833 Accession # P42702 | |
| RUO | |
| 3977 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 32953 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF R alpha, LIF R beta, LIF receptor, LIFR, LIF-R, LIFRa, SJS2, STWS, SWS | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LIF R alpha in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction