missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LIFR (P42702, Gln 45 - Ser 833) Partial Recombinant Protein expressed in HEK293 cells

Product Code. 16201990
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
This item is not returnable. View return policy

Product Code. 16201990

Brand: Abnova™ P3681.20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

This gene encodes a protein that belongs to the type I cytokine receptor family. This protein combines with a high-affinity converter subunit, gp130, to form a receptor complex that mediates the action of the leukemia inhibitory factor, a polyfunctional cytokine that is involved in cellular differentiation, proliferation and survival in the adult and the embryo. Mutations in this gene cause Schwartz-Jampel syndrome type 2, a disease belonging to the group of the bent-bone dysplasias. A translocation that involves the promoter of this gene, t(5;8)(p13;q12) with the pleiomorphic adenoma gene 1, is associated with salivary gland pleiomorphic adenoma, a common type of benign epithelial tumor of the salivary gland. Multiple splice variants encoding the same protein have been found for this gene. [provided by RefSeq]

Specifications

Accession Number P42702
For Use With (Application) SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 3977
Molecular Weight (g/mol) 125-135kDa
Name LIFR (Human) Recombinant Protein
Preparation Method Mammalian cell (HEK293) expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 20 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at 4°C for one month. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1 EU/μg)
Gene Alias CD118/FLJ98106/FLJ99923/LIF-R/SJS2/STWS/SWS
Common Name LIFR
Gene Symbol LIFR
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (HEK293) expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.