missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRA2/CD85h/ILT1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6364N-100UG
This item is not returnable.
View return policy
Description
ILT1/CD85h/LILRA2 Monoclonal specifically detects ILT1/CD85h/LILRA2 in Human samples. It is validated for Flow Cytometry.
Specifications
| ILT1/CD85h/LILRA2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8N149 | |
| LILRA2 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRA2/CD85h/ILT1 Pro17-Ser437 Accession # Q8N149 | |
| RUO | |
| 11027 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 600007 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD85 antigen-like family member H, CD85h, CD85h antigen, ILT1, ILT-1, ILT1CD85H, Leukocyte immunoglobulin-like receptor 7, leukocyte immunoglobulin-like receptor subfamily A member 2, leukocyte immunoglobulin-like receptor subfamily A member 2 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2, LIR7, LIR7LIR-7Immunoglobulin-like transcript 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LILRA2/CD85h/ILT1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT2, 3, 4, 5, rhLIR6, or rhLIR8 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction