missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRA6/CD85b Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8656R-100UG
This item is not returnable.
View return policy
Description
ILT8/CD85b/LILRA6 Monoclonal specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry.
Specifications
| ILT8/CD85b/LILRA6 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LILRA6/CD85b in direct ELISA. Detects HEK293 cells transfected with LILRA6/CD85b but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 921330 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6PI73 | |
| HEK293 human embryonic kidney cell line-derived recombinant human LILRA6/CD85b Met1-Asn447 Accession # Q6PI73 | |
| RUO | |
| 79168 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction