missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LINGO-1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB30861N-100UG
This item is not returnable.
View return policy
Description
LINGO-1 Monoclonal specifically detects LINGO-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| LINGO-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_116197 | |
| LINGO1 | |
| Mouse myeloma cell line NS0-derived recombinant human LINGO-1 Thr40-Thr556 Accession # NP_116197 | |
| RUO | |
| 84894 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 332237 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FLJ14594, LERN1, LERN1UNQ201, leucine rich repeat and Ig domain containing 1, leucine rich repeat neuronal 6A, Leucine-rich repeat and immunoglobilin domain-containing protein 1, Leucine-rich repeat neuronal protein 1, Leucine-rich repeat neuronal protein 6A, LINGO1, LRRN6A, LRRN6Aleucine-rich repeat neuronal 6A, MGC17422 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LINGO-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) LINGO-2 is observed and no cross-reactivity with rhLINGO-3 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction