missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LINGO-2 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB36791V-100UG
This item is not returnable.
View return policy
Description
LINGO-2 Monoclonal specifically detects LINGO-2 in Human samples. It is validated for Flow Cytometry.
Specifications
| LINGO-2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q7L985 | |
| LINGO2 | |
| Mouse myeloma cell line NS0-derived recombinant human LINGO-2 Cys28-Leu542 Accession # Q7L985 | |
| RUO | |
| 158038 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 382007 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| LERN3, LERN3FLJ31810, leucine rich repeat and Ig domain containing 2, leucine-rich repeat and immunoglobulin-like domain-containing nogoreceptor-interacting protein 2, Leucine-rich repeat neuronal protein 3, Leucine-rich repeat neuronal protein 6C, LINGO2, LRRN6C, LRRN6Cleucine rich repeat neuronal 6C | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LINGO-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LINGO-1 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction