Learn More
Novus Biologicals™ Human Liver Tissue Lysate (Adult Tumor)
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
The Human Liver Tissue Lysate (Adult) is derived from human.
Specifications
Specifications
| Tissue | Liver |
| Quantity | 0.1 mg |
| Host Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Target | Liver |
| Synonym | Liver, Liver lysate, liver lysates |