missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Livin Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1161
This item is not returnable.
View return policy
Description
Livin Polyclonal specifically detects Livin in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| Livin | |
| Polyclonal | |
| Unconjugated | |
| Q96CA5 | |
| BIRC7 | |
| E. coli-derived recombinant human Livin beta Met1-Gln280 Accession # Q96CA5 | |
| 100 μg | |
| Primary | |
| Detects human Livin in Western blots. Can detect recombinant and endogenous human Livin alpha (KIAP) and Livin beta (ML-IAP). | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, BIRC7, KIAP, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, ML-IAP, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 79444 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction