missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LOX-1/OLR1 PerCP-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB1798C
This item is not returnable.
View return policy
Description
LOX-1/OLR1 Monoclonal specifically detects LOX-1/OLR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| LOX-1/OLR1 | |
| Monoclonal | |
| PerCP | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4973 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 331212 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P78380 | |
| OLR1 | |
| Mouse myeloma cell line NS0-derived recombinant human LOX-1/OLR1 Ser61-Gln273 Accession # P78380 | |
| 100 Tests | |
| Primary | |
| Detects human LOX-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse LOX-1. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction