missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRIG1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems FAB8140G
This item is not returnable.
View return policy
Description
LRIG1 Polyclonal specifically detects LRIG1 in Human samples. It is validated for Flow Cytometry.Order Info
The product is shipped with polar packs.
Specifications
| LRIG1 | |
| Polyclonal | |
| Flow Cytometry 5 uL/10^6 cells | |
| Q96JA1 | |
| LRIG1 | |
| Human embryonic kidney cell line HEK293-derived recombinant human LRIG1 Ala35-Ser779 Accession # Q96JA1 | |
| 100 Tests | |
| Primary | |
| Detects human LRIG1 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse LRIG1 and and less 1% cross-reactivity with recombinant human LRIG3 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| D6Bwg0781e, Img, LIG1, LIG-1 | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| 26018 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction