missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRIG2 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB1941G-100UG
This item is not returnable.
View return policy
Description
LRIG2 Monoclonal specifically detects LRIG2 in Human samples. It is validated for Flow Cytometry.
Specifications
| LRIG2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
| KIAA0806, leucine-rich repeats and immunoglobulin-like domains 2, leucine-rich repeats and immunoglobulin-like domains protein 2, LIG2, UFS2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human LRIG-2 Gly41-Thr805 Accession # O94898 | |
| 100 μg | |
| 9860 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 990310 | |
| Flow Cytometry 0.25-1 μg/10^6 cells | |
| O94898 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human LRIG-2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction