missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRP-11 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8355-SP
This item is not returnable.
View return policy
Description
LRP-11 Polyclonal specifically detects LRP-11 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LRP-11 | |
| Polyclonal | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q86VZ4 | |
| LRP11 | |
| E. coli-derived recombinant human LRP-11 Ala38-Gly450 (Pro92Arg) Accession # Q86VZ4 | |
| 25 μg | |
| Primary | |
| Detects human LRP-11 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bA350J20.3, FLJ14735, low density lipoprotein receptor-related protein 11, low-density lipoprotein receptor-related protein 11, LRP11, LRP-11, MANSC3, MGC39092 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 84918 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction