missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome b5 type A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14923
This item is not returnable.
View return policy
Description
Cytochrome b5 type A Polyclonal specifically detects Cytochrome b5 type A in Human samples. It is validated for Western Blot.
Specifications
| Cytochrome b5 type A | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
| CYB5, cytochrome b-5, cytochrome b5 (microsomal), cytochrome b5 type A (microsomal), MCB5cytochrome b5, Microsomal cytochrome b5 type A, type 1 cyt-b5 | |
| Rabbit | |
| 15 kDa | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Stem Cell Markers | |
| 1528 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.28mg/mL | |
| Western Blot 1:500-1:3000 | |
| P00167 | |
| CYB5A | |
| Recombinant protein encompassing a sequence within the center region of human Cytochrome b5. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction