missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LY6G5B (aa 98-192) Control Fragment Recombinant Protein

Product Code. 30205038
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30205038 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30205038 Supplier Invitrogen™ Supplier No. RP100842

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61705 (PA5-61705. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LY6G5B belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NDX9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 58496
Name Human LY6G5B (aa 98-192) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C6orf19; G5B; LY6G5B; Lymphocyte antigen 6 complex locus protein G5b; lymphocyte antigen 6 complex, locus G5B; lymphocyte antigen 6 family member G5B; lymphocyte antigen-6 G5B splicing isoform 288; lymphocyte antigen-6 G5B splicing isoform 452; lymphocyte antigen-6 G5B splicing isoform 690; lymphocyte antigen-6 G5B splicing isoform 837
Common Name LY6G5B
Gene Symbol LY6G5B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CCQYDYCNSWSSPQLQSSLPEPHDRPLALPLSDSQIQWFYQALNLSLPLPNFHAGTEPDGLDPMVTLSLNLGLSFAELRRMYLFLNSSGLLVLPQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.