missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lymphotoxin-alpha/TNF-beta Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF-211-NA
This item is not returnable.
View return policy
Description
Lymphotoxin-alpha/TNF-beta Polyclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| Lymphotoxin-alpha/TNF-beta | |
| Polyclonal | |
| Unconjugated | |
| P01374 | |
| LTA | |
| E. coli-derived recombinant human Lymphotoxin-alpha/TNF-beta Leu35-Leu205 Accession # P01374 | |
| 100 μg | |
| Primary | |
| Detects human Lymphotoxin-alpha/TNF-beta in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) TNF-alpha , rhsTNF RI and rhsTNF RII is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization <200 ng/mL | |
| LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4049 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction