missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lymphotoxin beta R/TNFRSF3 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB629A
This item is not returnable.
View return policy
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Flow Cytometry.
Specifications
| Lymphotoxin beta R/TNFRSF3 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4055 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 71319 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P36941 | |
| LTBR | |
| Mouse myeloma cell line NS0-derived recombinant human Lymphotoxin beta R/TNFRSF3 Gln31-Met227 Accession # P36941 | |
| 100 Tests | |
| Primary | |
| Detects human Lymphotoxin beta R/TNFRSF3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, recombinant mouse (rm) Lymphotoxin beta R, rhNGF R, rhOPG, rhRELT, rhTAJ, rhTNF RI or rhTNF RII is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction