missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LYPD8 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9087R-100UG
This item is not returnable.
View return policy
Description
LYPD8 Monoclonal specifically detects LYPD8 in Human samples. It is validated for Flow Cytometry.
Specifications
| LYPD8 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human LYPD8 in direct ELISAs. Stains human LYPD8 transfectants but not irrelevant transfectants in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 961703 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q6UX82 | |
| Human embryonic kidney cell line HEK293-derived transfected with human LYPD8, Met1-Asn215, Accession # Q6UX82 | |
| 100 μg | |
| 646627 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction