missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysine (K)-specific Demethylase 4C/KDM4C Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6430-SP
This item is not returnable.
View return policy
Description
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Polyclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.14.11, EC 1.14.11.-, GASC1, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KDM4C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, Lysine (K)specific Demethylase 4C, lysine-specific demethylase 4C | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23081 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q9H3R0 | |
| KDM4C | |
| E. coli-derived recombinant human KDM4C Lys381-Lys588 Accession # Q9H3R0 | |
| 25 μg | |
| Primary | |
| Detects human KDM4C in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human KDM4A is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction